Anti-KIR3DS1 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: KIR3DS1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77822-558
- 77822-559
- Antigen symbol:
- KIR3DS1
- KIR3DS1
- Antigen name:
- Killer cell immunoglobulin-like receptor 3DS1 protein (198-335AA)
- Killer cell immunoglobulin-like receptor 3DS1 protein (198-335AA)
- Antigen synonyms:
- KIR3DS1|NKAT10Killer cell immunoglobulin-like receptor 3DS1|MHC class I NK cell receptor|Natural killer-associated transcript 10|NKAT-10
- KIR3DS1|NKAT10Killer cell immunoglobulin-like receptor 3DS1|MHC class I NK cell receptor|Natural killer-associated transcript 10|NKAT-10
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q14943
- Q14943
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together