Anti-phr Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: phr
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Synechococcus sp.
Specifications
- Catalog No:
- 77817-483
- 77817-484
- Antigen symbol:
- phr
- phr
- Antigen name:
- Recombinant Synechococcus sp. Deoxyribodipyrimidine photo-lyase protein (2-484AA)
- Recombinant Synechococcus sp. Deoxyribodipyrimidine photo-lyase protein (2-484AA)
- Antigen synonyms:
- phr|phrA|syc1392_c|Deoxyribodipyrimidine photo-lyase|EC 4.1.99.3|DNA photolyase|Photoreactivating enzyme
- phr|phrA|syc1392_c|Deoxyribodipyrimidine photo-lyase|EC 4.1.99.3|DNA photolyase|Photoreactivating enzyme
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Synechococcus sp.
- Synechococcus sp.
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P05327
- P05327
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together