Anti-SCARA5 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: SCARA5
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77798-715
- 77798-714
- Antigen symbol:
- SCARA5
- SCARA5
- Antigen name:
- Scavenger receptor class A member 5 protein (303-428AA)
- Scavenger receptor class A member 5 protein (303-428AA)
- Antigen synonyms:
- FLJ23907|MGC45780|NET33|SCAR5_HUMAN|SCARA 5|scara5|Scavenger receptor class A member 5|Scavenger receptor class A|member 5 (putative)|member 5|Scavenger receptor hlg|Tesr|Testis expressed scavenger receptor
- FLJ23907|MGC45780|NET33|SCAR5_HUMAN|SCARA 5|scara5|Scavenger receptor class A member 5|Scavenger receptor class A|member 5 (putative)|member 5|Scavenger receptor hlg|Tesr|Testis expressed scavenger receptor
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q6ZMJ2
- Q6ZMJ2
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together