Anti-TRIM26 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:200 to 1:500
Type:
Antigen: TRIM26
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77823-817
- 77823-816
- Antigen symbol:
- TRIM26
- TRIM26
- Antigen name:
- Tripartite motif-containing protein 26 protein (151-270AA)
- Tripartite motif-containing protein 26 protein (151-270AA)
- Antigen synonyms:
- Acid finger protein|AFP|CAM25547|RING finger protein 95|RNF95|TRI26_HUMAN|tripartite motif containing 26|Tripartite motif-containing protein 26|widely expressed acid zinc finger protein|Zinc finger protein 173|formerly|ZNF173
- Acid finger protein|AFP|CAM25547|RING finger protein 95|RNF95|TRI26_HUMAN|tripartite motif containing 26|Tripartite motif-containing protein 26|widely expressed acid zinc finger protein|Zinc finger protein 173|formerly|ZNF173
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q12899
- Q12899
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together