Anti-NPDC1 Rabbit Polyclonal Antibody
Catalog # 77813-250
Supplier: AFG Bioscience
New Product
Specifications
- Antigen name:Synthesized peptide derived from internal of NPDC1.
- Antigen symbol:NPDC1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:100 μl
- Form:Liquid
- Gene ID:Q9NQX5
- Antigen synonyms:Neural proliferation differentiation and control protein 1|NPDC-1
- Storage buffer:PBS, 150 mM NaCl, 0.02% sodium azide, 50% glycerol, pH 7.4.
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on Blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:3000
Type:
Antigen: NPDC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse