Anti-SPINT2 Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: SPINT2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77800-871
- 77800-872
- Antigen symbol:
- SPINT2
- SPINT2
- Antigen name:
- Kunitz-type protease inhibitor 2 protein (71-194AA)
- Kunitz-type protease inhibitor 2 protein (71-194AA)
- Antigen synonyms:
- AL024025|Bikunin, placental|C76321|DIAR3|FLJ45571|HAI-2|HAI2|Hepatocyte growth factor activator inhibitor type 2|Kop|Kunitz type protease inhibitor 2|Kunitz-type protease inhibitor 2|Kunitz-type serine protease inhibitor|MGC72638|PB|Placental bikunin|Serine peptidase inhibitor Kunitz type 2|Serine protease inhibitor, Kunitz type, 2|SPINT2|SPIT2_HUMAN
- AL024025|Bikunin, placental|C76321|DIAR3|FLJ45571|HAI-2|HAI2|Hepatocyte growth factor activator inhibitor type 2|Kop|Kunitz type protease inhibitor 2|Kunitz-type protease inhibitor 2|Kunitz-type serine protease inhibitor|MGC72638|PB|Placental bikunin|Serine peptidase inhibitor Kunitz type 2|Serine protease inhibitor, Kunitz type, 2|SPINT2|SPIT2_HUMAN
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O43291
- O43291
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together