Anti-CAMK1 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: CAMK1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77822-466
- 77822-465
- Antigen symbol:
- CAMK1
- CAMK1
- Antigen name:
- Calcium/calmodulin-dependent protein kinase type 1 protein (166-265AA)
- Calcium/calmodulin-dependent protein kinase type 1 protein (166-265AA)
- Antigen synonyms:
- Calcium / calmodulin dependent protein kinase 1|Calcium / calmodulin dependent protein kinase I|Calcium / calmodulin dependent protein kinase type 1|Calcium/calmodulin dependent protein kinase 1|Calcium/calmodulin dependent protein kinase I|Calcium/calmodulin dependent protein kinase type 1|Calcium/calmodulin-dependent protein kinase type 1|CaM K1|CaM KI|CaM kinase 1 alpha|CAM kinase 1|CaM kinase I alpha|CaM kinase I|CaM-KI|CaMK 1|CAMK I|CaMK1 alpha|CAMK1|CAMK1 PEN|CaMKI alpha|CaMKI-alpha|KCC1A_HUMAN|MGC120317|MGC120318
- Calcium / calmodulin dependent protein kinase 1|Calcium / calmodulin dependent protein kinase I|Calcium / calmodulin dependent protein kinase type 1|Calcium/calmodulin dependent protein kinase 1|Calcium/calmodulin dependent protein kinase I|Calcium/calmodulin dependent protein kinase type 1|Calcium/calmodulin-dependent protein kinase type 1|CaM K1|CaM KI|CaM kinase 1 alpha|CAM kinase 1|CaM kinase I alpha|CaM kinase I|CaM-KI|CaMK 1|CAMK I|CaMK1 alpha|CAMK1|CAMK1 PEN|CaMKI alpha|CaMKI-alpha|KCC1A_HUMAN|MGC120317|MGC120318
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q14012
- Q14012
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together