Anti-SLC2A4RG Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:1000 to 1:5000, WB: 1:200 to 1:1000
Type:
Antigen: SLC2A4RG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77820-356
- 77820-355
- Antigen symbol:
- SLC2A4RG
- SLC2A4RG
- Antigen name:
- Synthetic peptide of SLC2A4RG
- Synthetic peptide of SLC2A4RG
- Antigen synonyms:
- GEF|GLUT4 enhancer factor|HDBP 1|HDBP-1|HDBP1|Huntington disease gene regulatory region binding protein 1|Huntington disease gene regulatory region-binding protein 1|S2A4R_HUMAN|Si 1 2 19|Si 1 2|SLC2A4 regulator|SLC2A4RG|Zinc finger protein 2 5 Glut4 enhancer factor
- GEF|GLUT4 enhancer factor|HDBP 1|HDBP-1|HDBP1|Huntington disease gene regulatory region binding protein 1|Huntington disease gene regulatory region-binding protein 1|S2A4R_HUMAN|Si 1 2 19|Si 1 2|SLC2A4 regulator|SLC2A4RG|Zinc finger protein 2 5 Glut4 enhancer factor
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9NR83
- Q9NR83
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together