Order Entry
Canada
ContactUsLinkComponent
Anti-NTMT1 Rabbit Recombinant Polyclonal Antibody (FITC)
Anti-NTMT1 Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier:  AFG Bioscience
New Product
Certificates
undefined
Anti-NTMT1 Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier:  AFG Bioscience
New Product

Specifications

  • Catalog No:
  • 77840-312
  • 77840-313
  • Antigen symbol:
  • NTMT1
  • NTMT1
  • Antigen name:
  • N-terminal Xaa-Pro-Lys N-methyltransferase 1 protein (1-223AA)
  • N-terminal Xaa-Pro-Lys N-methyltransferase 1 protein (1-223AA)
  • Antigen synonyms:
  • AD 003| Alpha N-terminal protein methyltransferase 1A| C9orf32| Chromosome 9 open reading frame 32| Methyltransferase like 11A| Methyltransferase-like protein 11A| Mettl11a| N-terminal RCC1 methyltransferase| NRMT| NTM1A| NTM1A_HUMAN| NTMT1| X-Pro-Lys N-terminal protein methyltransferase 1A
  • AD 003| Alpha N-terminal protein methyltransferase 1A| C9orf32| Chromosome 9 open reading frame 32| Methyltransferase like 11A| Methyltransferase-like protein 11A| Mettl11a| N-terminal RCC1 methyltransferase| NRMT| NTM1A| NTM1A_HUMAN| NTMT1| X-Pro-Lys N-terminal protein methyltransferase 1A
  • Clonality:
  • Recombinant Polyclonal
  • Recombinant Polyclonal
  • Conjugation:
  • FITC (Fluorescein Isothiocyanate)
  • FITC (Fluorescein Isothiocyanate)
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • Q9BV86
  • Q9BV86
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • >95%, Protein G purified
  • >95%, Protein G purified
  • Storage buffer:
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • Storage temperature:
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Shipping temperature:
  • Shipped on Blue ice
  • Shipped on Blue ice

Specifications

Frequently Bought Together