Anti-IGF2BP1 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:1000 to 1:5000, WB: 1:500 to 1:2000, IHC: 1:50 to 1:200
Type:
Antigen: IGF2BP1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Specifications
- Catalog No:
- 77818-129
- 77818-128
- Antigen symbol:
- IGF2BP1
- IGF2BP1
- Antigen name:
- Synthetic peptide of IGF2BP1
- Synthetic peptide of IGF2BP1
- Antigen synonyms:
- Coding region determinant-binding protein| CRD BP| CRD-BP| CRDBP | IF2B1| IF2B1_HUMAN| IGF II mRNA binding protein 1 | IGF-II mRNA-binding protein 1| IGF2 mRNA binding protein 1| IGF2 mRNA-binding protein 1| IGF2BP1| IMP 1 | IMP-1| IMP1 | Insulin like growth factor 2 mRNA binding protein 1| Insulin-like growth factor 2 mRNA-binding protein 1| VICKZ family member 1| VICKZ1 | ZBP 1| ZBP-1| ZBP1 | Zip code binding protein 1| Zip code-binding protein 1| Zipcode binding protein 1| Zipcode-binding protein 1
- Coding region determinant-binding protein| CRD BP| CRD-BP| CRDBP | IF2B1| IF2B1_HUMAN| IGF II mRNA binding protein 1 | IGF-II mRNA-binding protein 1| IGF2 mRNA binding protein 1| IGF2 mRNA-binding protein 1| IGF2BP1| IMP 1 | IMP-1| IMP1 | Insulin like growth factor 2 mRNA binding protein 1| Insulin-like growth factor 2 mRNA-binding protein 1| VICKZ family member 1| VICKZ1 | ZBP 1| ZBP-1| ZBP1 | Zip code binding protein 1| Zip code-binding protein 1| Zipcode binding protein 1| Zipcode-binding protein 1
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse, Rat
- Human, Mouse, Rat
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9NZI8
- Q9NZI8
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together