Anti-SH3BGRL3 Rabbit Recombinant Polyclonal Antibody
Catalog # 77840-570
Supplier: AFG Bioscience
New Product
Specifications
- Antigen name:SH3 domain-binding glutamic acid-rich-like protein 3 protein (2-93AA)
- Antigen symbol:SH3BGRL3
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:Q9H299
- Antigen synonyms:epididymis secretory protein Li 297| HEL-S-297| SH3 domain binding glutamic acid rich protein like 3| SH3 domain-binding glutamic acid-rich-like protein 3| SH3 domain-binding protein 1| Sh3bgrl3| SH3BGRL3 like protein| SH3BP-1| SH3L3_HUMAN| TIP B1| TIP-B1| TNF inhibitory protein
- Storage buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on Blue ice
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:1000 to 1:5000, IHC: 1:20 to 1:200
Type:
Antigen: SH3BGRL3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human