Anti-ZEB2 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: ZEB2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77804-922
- 77804-923
- Antigen symbol:
- ZEB2
- ZEB2
- Antigen name:
- Zinc finger E-box-binding homeobox 2 protein (1050-1214AA)
- Zinc finger E-box-binding homeobox 2 protein (1050-1214AA)
- Antigen synonyms:
- FLJ42816|HSPC082|KIAA0569|SIP 1|SIP1|Smad Interacting Protein 1|Smad-interacting protein 1|SMADIP 1|SMADIP1|ZEB 2|Zeb2|ZEB2_HUMAN|Zfhx1b|ZFHX1B protein|Zfx1b|Zinc finger E box binding protein 2|Zinc finger E-box-binding homeobox 2|Zinc finger homeobox 1b|zinc finger homeobox protein 1|Zinc finger homeobox protein 1b
- FLJ42816|HSPC082|KIAA0569|SIP 1|SIP1|Smad Interacting Protein 1|Smad-interacting protein 1|SMADIP 1|SMADIP1|ZEB 2|Zeb2|ZEB2_HUMAN|Zfhx1b|ZFHX1B protein|Zfx1b|Zinc finger E box binding protein 2|Zinc finger E-box-binding homeobox 2|Zinc finger homeobox 1b|zinc finger homeobox protein 1|Zinc finger homeobox protein 1b
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O60315
- O60315
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage temperature:
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together