Anti-PELO Rabbit Recombinant Polyclonal Antibody (HRP)
New Product
Type:
Antigen: PELO
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77835-212
- 77835-213
- Antigen symbol:
- PELO
- PELO
- Antigen name:
- Protein pelota homolog protein (1-385AA)
- Protein pelota homolog protein (1-385AA)
- Antigen synonyms:
- CGI 17|CGI17|pelo|PELO_HUMAN|pelota (Drosophila) homolog|pelota homolog|PRO1770|Protein pelota homolog
- CGI 17|CGI17|pelo|PELO_HUMAN|pelota (Drosophila) homolog|pelota homolog|PRO1770|Protein pelota homolog
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9BRX2
- Q9BRX2
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together