Anti-PDXDC1 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: PDXDC1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77831-149
- 77831-150
- Antigen symbol:
- PDXDC1
- PDXDC1
- Antigen name:
- Pyridoxal-dependent decarboxylase domain-containing protein 1 protein (681-780AA)
- Pyridoxal-dependent decarboxylase domain-containing protein 1 protein (681-780AA)
- Antigen synonyms:
- 2210010A19Rik| AA415817| KIAA0251| Kiaa0251 hp| LP8165| MGC189517| MGC8182| PDXD1_HUMAN| PDXDC 1| Pdxdc1| Pyridoxal dependent decarboxylase domain containing 1| Pyridoxal-dependent decarboxylase domain-containing protein 1| rCG_49895| RGD1562597
- 2210010A19Rik| AA415817| KIAA0251| Kiaa0251 hp| LP8165| MGC189517| MGC8182| PDXD1_HUMAN| PDXDC 1| Pdxdc1| Pyridoxal dependent decarboxylase domain containing 1| Pyridoxal-dependent decarboxylase domain-containing protein 1| rCG_49895| RGD1562597
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q6P996
- Q6P996
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together