Anti-WDR77 Rabbit Recombinant Polyclonal Antibody (HRP)
New Product
Type:
Antigen: WDR77
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77834-674
- 77834-673
- Antigen symbol:
- WDR77
- WDR77
- Antigen name:
- Methylosome protein 50 protein (1-312AA)
- Methylosome protein 50 protein (1-312AA)
- Antigen synonyms:
- 2610312E17Rik|Androgen receptor cofactor p44|C79984|HKMT1069|MEP 50|MEP-50|MEP50|MEP50_HUMAN|Methylosome protein 50|MGC2722|Nbla10071|p44|p44/Mep50|RGD1310479 |RP11 552M11.3|WD repeat containing protein 77|WD repeat domain 77 |WD repeat-containing protein 77|WDR77
- 2610312E17Rik|Androgen receptor cofactor p44|C79984|HKMT1069|MEP 50|MEP-50|MEP50|MEP50_HUMAN|Methylosome protein 50|MGC2722|Nbla10071|p44|p44/Mep50|RGD1310479 |RP11 552M11.3|WD repeat containing protein 77|WD repeat domain 77 |WD repeat-containing protein 77|WDR77
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9BQA1
- Q9BQA1
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, protein G purified
- >95%, protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together