Anti-SMAD9 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: WB: 1:500 to 1:2000, IHC: 1:20 to 1:200
Type:
Antigen: SMAD9
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77800-274
- 77800-275
- Antigen symbol:
- SMAD9
- SMAD9
- Antigen name:
- Mothers against decapentaplegic homolog 9 protein (1-300AA)
- Mothers against decapentaplegic homolog 9 protein (1-300AA)
- Antigen synonyms:
- MAD homolog 9|Madh6|Mothers against decapentaplegic|Mothers against decapentaplegic homolog 9|Mothers against DPP homolog 9|SMAD 9|SMAD family member 9|SMAD, mothers against DPP homolog 9 (Drosophila)|SMAD8A|SMAD8B|SMAD9_HUMAN
- MAD homolog 9|Madh6|Mothers against decapentaplegic|Mothers against decapentaplegic homolog 9|Mothers against DPP homolog 9|SMAD 9|SMAD family member 9|SMAD, mothers against DPP homolog 9 (Drosophila)|SMAD8A|SMAD8B|SMAD9_HUMAN
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O15198
- O15198
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together