Anti-PTGR2 Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: PTGR2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77796-014
- 77796-013
- Antigen symbol:
- PTGR2
- PTGR2
- Antigen name:
- Prostaglandin reductase 2 protein (206-282AA)
- Prostaglandin reductase 2 protein (206-282AA)
- Antigen synonyms:
- 15 oxoprostaglandin 13 reductase|15 oxoprostaglandin delta 13 reductase|15-oxoprostaglandin 13-reductase|DKFZp686P10120|FLJ39091|FLJ99229|PGR 2|PGR2|PRG-2|Prostaglandin reductase 2|PTGR 2|PTGR2|PTGR2_HUMAN|Zinc binding alcohol dehydrogenase domain containing 1|Zinc binding alcohol dehydrogenase domain containing protein 1|Zinc-binding alcohol dehydrogenase domain-containing protein 1
- 15 oxoprostaglandin 13 reductase|15 oxoprostaglandin delta 13 reductase|15-oxoprostaglandin 13-reductase|DKFZp686P10120|FLJ39091|FLJ99229|PGR 2|PGR2|PRG-2|Prostaglandin reductase 2|PTGR 2|PTGR2|PTGR2_HUMAN|Zinc binding alcohol dehydrogenase domain containing 1|Zinc binding alcohol dehydrogenase domain containing protein 1|Zinc-binding alcohol dehydrogenase domain-containing protein 1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8N8N7
- Q8N8N7
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together