Anti-LRRK1 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: LRRK1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77826-027
- 77826-026
- Antigen symbol:
- LRRK1
- LRRK1
- Antigen name:
- Leucine-rich repeat serine/threonine-protein kinase 1 protein (1-68AA)
- Leucine-rich repeat serine/threonine-protein kinase 1 protein (1-68AA)
- Antigen synonyms:
- KIAA1790|Leucine rich repeat serine/threonine protein kinase 1|Leucine-rich repeat serine/threonine-protein kinase 1|LRRK 1|Lrrk1|LRRK1_HUMAN|RIPK6|Roco1
- KIAA1790|Leucine rich repeat serine/threonine protein kinase 1|Leucine-rich repeat serine/threonine-protein kinase 1|LRRK 1|Lrrk1|LRRK1_HUMAN|RIPK6|Roco1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q38SD2
- Q38SD2
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together