Anti-pepF Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: pepF
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mycoplasma pneumoniae
Specifications
- Catalog No:
- 77816-576
- 77816-577
- Antigen symbol:
- pepF
- pepF
- Antigen name:
- Recombinant Mycoplasma pneumoniae Oligoendopeptidase F homolog protein (1-210AA)
- Recombinant Mycoplasma pneumoniae Oligoendopeptidase F homolog protein (1-210AA)
- Antigen synonyms:
- pepF| MPN_197| MP634Oligoendopeptidase F homolog| EC 3.4.24.-
- pepF| MPN_197| MP634Oligoendopeptidase F homolog| EC 3.4.24.-
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Mycoplasma pneumoniae
- Mycoplasma pneumoniae
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P54125
- P54125
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together