Anti-STAC Rabbit Recombinant Antibody
Catalog # 77834-694
Supplier: AFG Bioscience
New Product
Specifications
- Antigen name:SH3 and cysteine-rich domain-containing protein (1-119AA)
- Antigen symbol:STAC
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:Q99469
- Antigen synonyms:FLJ32331|SH3 and cysteine rich domain|SH3 and cysteine rich domain containing protein|SH3 and cysteine-rich domain-containing protein|SRC homology 3 and cysteine rich domain protein|Src homology 3 and cysteine-rich domain-containing protein|Src homology three (SH3) and cysteine rich domain|STAC 1|STAC|STAC_HUMAN|STAC1
- Storage buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:5000, IF: 1:50 to 1:200
Type:
Antigen: STAC
Clonality: Recombinant
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse