Anti-SLC35D3 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: SLC35D3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77800-046
- 77800-045
- Antigen symbol:
- SLC35D3
- SLC35D3
- Antigen name:
- Solute carrier family 35 member D3 protein (304-416AA)
- Solute carrier family 35 member D3 protein (304-416AA)
- Antigen synonyms:
- bA55K22.3|Frc fringe like 1|FRCL1|Fringe connection like protein 1|Fringe connection-like protein 1|MGC102873|S35D3_HUMAN|SLC35D3|Solute carrier family 35 member D3
- bA55K22.3|Frc fringe like 1|FRCL1|Fringe connection like protein 1|Fringe connection-like protein 1|MGC102873|S35D3_HUMAN|SLC35D3|Solute carrier family 35 member D3
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q5M8T2
- Q5M8T2
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, protein G purified
- >95%, protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together