Anti-ELOVL1 Rabbit Recombinant Polyclonal Antibody
New Product
Recommended dilution: IHC: 1:20 to 1:200, IF: 1:50 to 1:200
Type:
Antigen: ELOVL1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77842-920
- 77842-919
- Antigen symbol:
- ELOVL1
- ELOVL1
- Antigen name:
- Elongation of very long chain fatty acids protein 1 protein (82-175AA)
- Elongation of very long chain fatty acids protein 1 protein (82-175AA)
- Antigen synonyms:
- 3 keto acyl CoA synthase ELOVL1|3-keto acyl-CoA synthase ELOVL1|CGI 88|Elongation of very long chain fatty acids (FEN1/Elo2 SUR4/Elo3 yeast) like 1|Elongation of very long chain fatty acids like 1|Elongation of very long chain fatty acids protein 1|ELOV1_HUMAN|ELOVL 1|ELOVL FA elongase 1|ELOVL fatty acid elongase 1|ELOVL1|Ssc1|mouse|homolog of|very long chain 3 oxoacyl CoA synthase 1|Very-long-chain 3-oxoacyl-CoA synthase 1
- 3 keto acyl CoA synthase ELOVL1|3-keto acyl-CoA synthase ELOVL1|CGI 88|Elongation of very long chain fatty acids (FEN1/Elo2 SUR4/Elo3 yeast) like 1|Elongation of very long chain fatty acids like 1|Elongation of very long chain fatty acids protein 1|ELOV1_HUMAN|ELOVL 1|ELOVL FA elongase 1|ELOVL fatty acid elongase 1|ELOVL1|Ssc1|mouse|homolog of|very long chain 3 oxoacyl CoA synthase 1|Very-long-chain 3-oxoacyl-CoA synthase 1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9BW60
- Q9BW60
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together