Anti-NPRL3 Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: NPRL3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77823-836
- 77823-835
- Antigen symbol:
- NPRL3
- NPRL3
- Antigen name:
- GATOR complex protein NPRL3 protein (349-482AA)
- GATOR complex protein NPRL3 protein (349-482AA)
- Antigen synonyms:
- otein|Alpha-globin regulatory element-containing gene protein|C16ORF35|CGTHBA|Conserved gene telomeric to alpha globin cluster|HS 40|MARE|Nitrogen permease regulator 3-like protein|Nitrogen permease regulator-like 3 (S. cerevisiae)|NPR3|NPRL3|NPRL3_HUMAN|Protein CGTHBA|RMD11
- otein|Alpha-globin regulatory element-containing gene protein|C16ORF35|CGTHBA|Conserved gene telomeric to alpha globin cluster|HS 40|MARE|Nitrogen permease regulator 3-like protein|Nitrogen permease regulator-like 3 (S. cerevisiae)|NPR3|NPRL3|NPRL3_HUMAN|Protein CGTHBA|RMD11
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q12980
- Q12980
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together