Anti-CHRDL1 Rabbit Recombinant Polyclonal Antibody
Catalog # 77836-831
Supplier: AFG Bioscience
New Product
Specifications
- Antigen name:Chordin-like protein 1 protein (92-237AA)
- Antigen symbol:CHRDL1
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:Q9BU40
- Antigen synonyms:CHL|chordin like 1|Chordin-like protein 1|CHRDL1|CRDL1_HUMAN|dA141H5.1|neuralin 1|Neuralin-1|Neurogenesin-1|NRLN1|Ventroptin|VOPT
- Storage buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:5000, IHC: 1:200 to 1:500
Type:
Antigen: CHRDL1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human