Anti-UBE2Z Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: WB: 1:1000 to 1:5000, IHC: 1:20 to 1:200
Type:
Antigen: UBE2Z
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
Specifications
- Catalog No:
- 77839-505
- 77839-504
- Antigen symbol:
- UBE2Z
- UBE2Z
- Antigen name:
- Ubiquitin-conjugating enzyme E2 Z protein (115-354AA)
- Ubiquitin-conjugating enzyme E2 Z protein (115-354AA)
- Antigen synonyms:
- FLJ13855|GC17P044341|HOYS7|Uba6 specific E2 conjugating enzyme 1|UBA6 specific enzyme E2|Uba6-specific E2 conjugating enzyme 1|UBE2Z|UBE2Z_HUMAN|Ubiquitin carrier protein Z|Ubiquitin conjugating enzyme E2 Z|Ubiquitin conjugating enzyme E2Z|Ubiquitin protein ligase Z|Ubiquitin-conjugating enzyme E2 Z|Ubiquitin-protein ligase Z|Use1
- FLJ13855|GC17P044341|HOYS7|Uba6 specific E2 conjugating enzyme 1|UBA6 specific enzyme E2|Uba6-specific E2 conjugating enzyme 1|UBE2Z|UBE2Z_HUMAN|Ubiquitin carrier protein Z|Ubiquitin conjugating enzyme E2 Z|Ubiquitin conjugating enzyme E2Z|Ubiquitin protein ligase Z|Ubiquitin-conjugating enzyme E2 Z|Ubiquitin-protein ligase Z|Use1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9H832
- Q9H832
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.02% Sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% Sodium azide, 50% glycerol, pH 7.3
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together