Anti-TMED3 Rabbit Polyclonal Antibody
Catalog # 77802-800
Supplier: AFG Bioscience
New Product
Specifications
- Antigen name:UNQ5357/PRO1078
- Antigen symbol:TMED3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q9Y3Q3
- Antigen synonyms:C15orf22|Integral type I protein|Membrane protein p24B|p24 family protein gamma 4|p24 family protein gamma-4|P24B|p24g4|p24gamma4|p26|Testis tissue sperm binding protein Li 48e|TMED3_HUMAN|Transmembrane emp24 domain containing protein 3|Transmembrane emp24 domain-containing protein 3|Transmembrane p24 trafficking protein 3|UNQ5357/PRO1078
- Storage buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: TMED3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat