Anti-RAB4A Rabbit Recombinant Polyclonal Antibody
Fournisseur: AFG Bioscience
New Product
Certificats
À propos de cet article
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: RAB4A
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Spécifications
- Catalog No:
- 77796-438
- 77796-439
- Antigen symbol:
- RAB4A
- RAB4A
- Antigen name:
- Ras-related protein Rab-4A protein (1-218AA)
- Ras-related protein Rab-4A protein (1-218AA)
- Antigen synonyms:
- HRES 1 / RAB4|Oncogene RAB4|Rab 4|RAB 4A|RAB4 member RAS oncogene family|Rab4a|RAB4A member RAS oncogene family|RAB4A_HUMAN|Ras related protein Rab 4A|Ras related protein Rab4A|Ras-related protein Rab-4A
- HRES 1 / RAB4|Oncogene RAB4|Rab 4|RAB 4A|RAB4 member RAS oncogene family|Rab4a|RAB4A member RAS oncogene family|RAB4A_HUMAN|Ras related protein Rab 4A|Ras related protein Rab4A|Ras-related protein Rab-4A
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P20338
- P20338
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Spécifications
Frequently Bought Together