Anti-CACNG4 Rabbit Polyclonal Antibody
77812-566
New Product
- Antigen name:Synthetic Peptide
- Antigen symbol:CACNG4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:Q9UBN1
- Antigen synonyms:CACNG4Voltage-dependent calcium channel gamma-4 subunit|Neuronal voltage-gated calcium channel gamma-4 subunit|Transmembrane AMPAR regulatory protein gamma-4|TARP gamma-4
- Storage buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.4
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Purification:Affinity purified
- Packaging:Vial
Recommended dilution: WB:1:1000 to 2000, IHC:1:200 to 500
Type:
Antigen: CACNG4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat