Anti-SYT1 Rabbit Polyclonal Antibody
77801-745
New Product
- Antigen name:SytI
- Antigen symbol:SYT1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western blot:Yes
- Size:100 μl
- Form:Liquid
- Gene ID:P21579
- Antigen synonyms:DKFZp781D2042|FLJ42519|p65|SVP65|Synaptotagmin 2|Synaptotagmin I|Synaptotagmin II|Synaptotagmin-1|SYT|SYT1_HUMAN|Syt2
- Storage buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3.
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on Blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Type:
Antigen: SYT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat