Anti-RBSN Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: RBSN
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77841-971
- 77841-970
- Antigen symbol:
- RBSN
- RBSN
- Antigen name:
- Rabenosyn-5 protein (201-500AA)
- Rabenosyn-5 protein (201-500AA)
- Antigen synonyms:
- 110 kDa protein|FYVE finger containing Rab5 effector protein Rabenosyn 5|FYVE finger-containing Rab5 effector protein rabenosyn-5|Rabenosyn-5|RBNS5_HUMAN|Zfyve20|zinc finger FYVE domain containing 20|Zinc finger FYVE domain-containing protein 20
- 110 kDa protein|FYVE finger containing Rab5 effector protein Rabenosyn 5|FYVE finger-containing Rab5 effector protein rabenosyn-5|Rabenosyn-5|RBNS5_HUMAN|Zfyve20|zinc finger FYVE domain containing 20|Zinc finger FYVE domain-containing protein 20
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9H1K0
- Q9H1K0
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together