Anti-SCNN1B Rabbit Recombinant Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: SCNN1B
Clonality: Recombinant
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77798-827
- 77798-826
- Antigen symbol:
- SCNN1B
- SCNN1B
- Antigen name:
- Amiloride-sensitive sodium channel subunit beta protein (554-640AA)
- Amiloride-sensitive sodium channel subunit beta protein (554-640AA)
- Antigen synonyms:
- Amiloride sensitive sodium channel subunit beta|Amiloride-sensitive sodium channel subunit beta|Beta NaCH|Beta-ENaC|Beta-NaCH|ENaC beta|ENaCB|Epithelial Na(+) channel subunit beta|Epithelial Na+ channel beta subunit|Epithelial Na+ channel subunit beta|Epithelial sodium channel beta 2 subunit|Epithelial sodium channel beta 3 subunit|Nonvoltage gated sodium channel 1 beta subunit|Nonvoltage gated sodium channel 1 subunit beta|Nonvoltage-gated sodium channel 1 subunit beta|SCNEB|SCNN 1B|SCNN1B|SCNNB_HUMAN|Sodium channel nonvoltage gated 1 beta (Liddle syndrome)|Sodium channel nonvoltage gated 1 beta
- Amiloride sensitive sodium channel subunit beta|Amiloride-sensitive sodium channel subunit beta|Beta NaCH|Beta-ENaC|Beta-NaCH|ENaC beta|ENaCB|Epithelial Na(+) channel subunit beta|Epithelial Na+ channel beta subunit|Epithelial Na+ channel subunit beta|Epithelial sodium channel beta 2 subunit|Epithelial sodium channel beta 3 subunit|Nonvoltage gated sodium channel 1 beta subunit|Nonvoltage gated sodium channel 1 subunit beta|Nonvoltage-gated sodium channel 1 subunit beta|SCNEB|SCNN 1B|SCNN1B|SCNNB_HUMAN|Sodium channel nonvoltage gated 1 beta (Liddle syndrome)|Sodium channel nonvoltage gated 1 beta
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P51168
- P51168
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together