Anti-RPS2 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: RPS2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77798-102
- 77798-103
- Antigen symbol:
- RPS2
- RPS2
- Antigen name:
- 40S ribosomal protein S2 protein (2-61AA)
- 40S ribosomal protein S2 protein (2-61AA)
- Antigen synonyms:
- 40S ribosomal protein S2|40S ribosomal protein S4|LLREP3|LLRep3 protein|MGC102851|MGC117344|MGC117345|OK/KNS-cl.6|Protein LLRep3|Ribosomal protein S2|rps2|RPS4|RS2|RS2_HUMAN|S2|S4
- 40S ribosomal protein S2|40S ribosomal protein S4|LLREP3|LLRep3 protein|MGC102851|MGC117344|MGC117345|OK/KNS-cl.6|Protein LLRep3|Ribosomal protein S2|rps2|RPS4|RS2|RS2_HUMAN|S2|S4
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P15880
- P15880
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together