Anti-GNAT3 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:2000 to 1:5000, WB: 1:500 to 1:2000, IHC: 1:25 to 1:100
Type:
Antigen: GNAT3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Specifications
- Catalog No:
- 77827-143
- 77827-144
- Antigen symbol:
- GNAT3
- GNAT3
- Antigen name:
- Synthetic peptide of GNAT3
- Synthetic peptide of GNAT3
- Antigen synonyms:
- GDCA|Ggust|Gnat 3|GNAT3|GNAT3_HUMAN|Gtn|Guanine nucleotide binding protein alpha transducing 3|Guanine nucleotide binding protein G(t) subunit alpha 3|Guanine nucleotide-binding protein G(t) subunit alpha-3|Gustducin alpha 3|Gustducin alpha 3 chain|Gustducin alpha-3 chain
- GDCA|Ggust|Gnat 3|GNAT3|GNAT3_HUMAN|Gtn|Guanine nucleotide binding protein alpha transducing 3|Guanine nucleotide binding protein G(t) subunit alpha 3|Guanine nucleotide-binding protein G(t) subunit alpha-3|Gustducin alpha 3|Gustducin alpha 3 chain|Gustducin alpha-3 chain
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse, Rat
- Human, Mouse, Rat
- Host:
- Rabbit
- Rabbit
- Gene ID:
- A8MTJ3
- A8MTJ3
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage temperature:
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together