Anti-CXCL6 Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: CXCL6
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77809-513
- 77809-514
- Antigen symbol:
- CXCL6
- CXCL6
- Antigen name:
- C-X-C motif chemokine 6 protein (38-114AA)
- C-X-C motif chemokine 6 protein (38-114AA)
- Antigen synonyms:
- C X C motif chemokine 6|Chemokine (C X C motif) ligand 6|Chemokine alpha 3|CKA 3|CKA-3|CKA3|CXCL 6|CXCL6|CXCL6_HUMAN|GCP 2|GCP-2|Granulocyte chemotactic protein 2|N-processed variant 3|SCYB6|Small inducible cytokine B6|Small inducible cytokine subfamily B (Cys X Cys) member 6|Small inducible cytokine subfamily B (Cys X Cys) member b|Small-inducible cytokine B6
- C X C motif chemokine 6|Chemokine (C X C motif) ligand 6|Chemokine alpha 3|CKA 3|CKA-3|CKA3|CXCL 6|CXCL6|CXCL6_HUMAN|GCP 2|GCP-2|Granulocyte chemotactic protein 2|N-processed variant 3|SCYB6|Small inducible cytokine B6|Small inducible cytokine subfamily B (Cys X Cys) member 6|Small inducible cytokine subfamily B (Cys X Cys) member b|Small-inducible cytokine B6
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P80162
- P80162
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together