Anti-TNFSF14 Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: TNFSF14
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 77810-082
- 77810-081
- Antigen symbol:
- TNFSF14
- TNFSF14
- Antigen name:
- Tumor necrosis factor ligand superfamily member 14 protein (1-239AA)
- Tumor necrosis factor ligand superfamily member 14 protein (1-239AA)
- Antigen synonyms:
- CD 258|CD258|CD258 antigen|Delta transmembrane LIGHT|Herpes virus entry mediator ligand|Herpesvirus entry mediator A|Herpesvirus entry mediator ligand|herpesvirus entry mediator-ligand|HVEM L|HVEM-L|HVEML|Ligand for herpesvirus entry mediator|LIGHT|LTg|soluble form|TNF14|TNF14_HUMAN|TNFSF 14|Tnfsf14|TNFSF14 protein|TR 2|TR2|Tumor necrosis factor (ligand) superfamily|member 14|Tumor necrosis factor ligand superfamily member 14|Tumor necrosis factor ligand superfamily|Tumor necrosis factor receptor like 2|tumor necrosis factor receptor-like 2|Tumor necrosis factor superfamily member 14|Tumor necrosis factor superfamily member LIGHT
- CD 258|CD258|CD258 antigen|Delta transmembrane LIGHT|Herpes virus entry mediator ligand|Herpesvirus entry mediator A|Herpesvirus entry mediator ligand|herpesvirus entry mediator-ligand|HVEM L|HVEM-L|HVEML|Ligand for herpesvirus entry mediator|LIGHT|LTg|soluble form|TNF14|TNF14_HUMAN|TNFSF 14|Tnfsf14|TNFSF14 protein|TR 2|TR2|Tumor necrosis factor (ligand) superfamily|member 14|Tumor necrosis factor ligand superfamily member 14|Tumor necrosis factor ligand superfamily|Tumor necrosis factor receptor like 2|tumor necrosis factor receptor-like 2|Tumor necrosis factor superfamily member 14|Tumor necrosis factor superfamily member LIGHT
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O43557
- O43557
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, protein G purified
- >95%, protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together