Anti-MTMR2 Rabbit Recombinant Polyclonal Antibody
77821-889
New Product
- Antigen name:Myotubularin-related protein 2 protein (1-173AA)
- Antigen symbol:MTMR2
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:Q13614
- Antigen synonyms:CMT4B| CMT4B1| KIAA1073| MTMR2| MTMR2_HUMAN| Myotubularin related protein 2| Myotubularin-related protein 2| OTTHUMP00000204445| OTTHUMP00000204446| OTTHUMP00000204447| OTTHUMP00000204448| Phosphatidylinositol 3 phosphate phosphatase| Phosphatidylinositol 3,5 bisphosphate 3 phosphatase
- Storage buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on Blue ice
- Purification:>95%, Protein G purified
- Packaging:Vial
Recommended dilution: WB: 1:500 to 1:5000, IHC: 1:300 to 1:500, IF: 1:50 to 1:200
Type:
Antigen: MTMR2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human