Anti-PAFAH2 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: PAFAH2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77834-406
- 77834-405
- Antigen symbol:
- PAFAH2
- PAFAH2
- Antigen name:
- Platelet-activating factor acetylhydrolase 2, cytoplasmic protein (1-206AA)
- Platelet-activating factor acetylhydrolase 2, cytoplasmic protein (1-206AA)
- Antigen synonyms:
- PAFAH2Platelet-activating factor acetylhydrolase 2|cytoplasmic|EC 3.1.1.47|Serine-dependent phospholipase A2|SD-PLA2|hSD-PLA2
- PAFAH2Platelet-activating factor acetylhydrolase 2|cytoplasmic|EC 3.1.1.47|Serine-dependent phospholipase A2|SD-PLA2|hSD-PLA2
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q99487
- Q99487
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together