Anti-ADIPOQ Rabbit Recombinant Polyclonal Antibody (FITC)
New Product
Type:
Antigen: ADIPOQ
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77812-336
- 77812-335
- Antigen symbol:
- ADIPOQ
- ADIPOQ
- Antigen name:
- Adiponectin protein (19-244AA)
- Adiponectin protein (19-244AA)
- Antigen synonyms:
- 30 kDa adipocyte complement related protein|30 kDa adipocyte complement-related protein|ACDC|Acrp 30|ACRP30|ADIPO_HUMAN|Adipocyte|Adipocyte C1q and collagen domain containing protein|Adipocyte complement related 30 kDa protein|Adipocyte complement related protein of 30 kDa|Adipocyte complement-related 30 kDa protein|adipocyte-specific secretory protein|Adiponectin|Adiponectin precursor|C1Q and collagen domain containing|Adipose most abundant gene transcript 1|Adipose most abundant gene transcript 1 protein|Adipose specific collagen like factor|ADIPQTL1|ADPN|APM 1|apM-1|APM1|C1q and collagen domain-containing protein|GBP 28|GBP28|Gelatin binding protein|Gelatin binding protein 28|gelatin-binding protein 28|Gelatin-binding protein|OTTHUMP00000210047
- 30 kDa adipocyte complement related protein|30 kDa adipocyte complement-related protein|ACDC|Acrp 30|ACRP30|ADIPO_HUMAN|Adipocyte|Adipocyte C1q and collagen domain containing protein|Adipocyte complement related 30 kDa protein|Adipocyte complement related protein of 30 kDa|Adipocyte complement-related 30 kDa protein|adipocyte-specific secretory protein|Adiponectin|Adiponectin precursor|C1Q and collagen domain containing|Adipose most abundant gene transcript 1|Adipose most abundant gene transcript 1 protein|Adipose specific collagen like factor|ADIPQTL1|ADPN|APM 1|apM-1|APM1|C1q and collagen domain-containing protein|GBP 28|GBP28|Gelatin binding protein|Gelatin binding protein 28|gelatin-binding protein 28|Gelatin-binding protein|OTTHUMP00000210047
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q15848
- Q15848
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together