Anti-RBP5 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: RBP5
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotinylated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77796-872
- 77796-871
- Antigen symbol:
- RBP5
- RBP5
- Antigen name:
- Retinol-binding protein 5 protein (1-135AA)
- Retinol-binding protein 5 protein (1-135AA)
- Antigen synonyms:
- Cellular retinol-binding protein III| CRBP III| CRBP-III| CRBP3| CRBPIII| HRBPiso| OTTHUMP00000238163| Putative cellular retinol binding protein CRBP III| RBP5| RET5_HUMAN| Retinol binding protein| Retinol binding protein 5| cellular| Retinol-binding protein 5
- Cellular retinol-binding protein III| CRBP III| CRBP-III| CRBP3| CRBPIII| HRBPiso| OTTHUMP00000238163| Putative cellular retinol binding protein CRBP III| RBP5| RET5_HUMAN| Retinol binding protein| Retinol binding protein 5| cellular| Retinol-binding protein 5
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P82980
- P82980
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together