Anti-MAPKAPK2 Rabbit Polyclonal Antibody
77810-526
New Product
- Antigen name:Synthesized peptide derived from MAPKAPK-2 around the phosphorylation site of T222.
- Antigen symbol:MAPKAPK2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat,Monkey
- Western blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:P49137
- Antigen synonyms:MAP kinase activated protein Kinase 2|MAP kinase-activated protein kinase 2|MAPK activated protein kinase 2|MAPK-activated protein kinase 2|MAPK2_HUMAN|MAPKAP K2|MAPKAP kinase 2|MAPKAPK 2|MAPKAPK-2|MAPKAPK2|Mitogen activated protein kinase activated protein kinase 2|MK 2|MK2
- Storage buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4.
- Storage temperature:Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Recommended dilution: WB: 1:500 to 1:2000, IHC: 1:100 to 1:300, ELISA: 1:20000
Type:
Antigen: MAPKAPK2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Monkey