Anti-USO1 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: USO1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77804-099
- 77804-098
- Antigen symbol:
- USO1
- USO1
- Antigen name:
- General vesicular transport factor p115 protein (591-733AA)
- General vesicular transport factor p115 protein (591-733AA)
- Antigen synonyms:
- General vesicular transport factor|General vesicular transport factor p115|P115|Protein USO1 homolog|TAP|Transcytosis associated protein|Transcytosis-associated protein|Uso1|USO1_HUMAN|VDP|Vesicle docking protein|Vesicle docking protein p115|Vesicle-docking protein
- General vesicular transport factor|General vesicular transport factor p115|P115|Protein USO1 homolog|TAP|Transcytosis associated protein|Transcytosis-associated protein|Uso1|USO1_HUMAN|VDP|Vesicle docking protein|Vesicle docking protein p115|Vesicle-docking protein
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O60763
- O60763
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together