Anti-SMARCA4 Rabbit Recombinant Polyclonal Antibody
New Product
Recommended dilution: IHC: 1:20 to 1:200, IF: 1:50 to 1:200
Type:
Antigen: SMARCA4
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77800-298
- 77800-297
- Antigen symbol:
- SMARCA4
- SMARCA4
- Antigen name:
- Transcription activator BRG1 protein (808-1092AA)
- Transcription activator BRG1 protein (808-1092AA)
- Antigen synonyms:
- ATP dependent helicase SMARCA4|ATP-dependent helicase SMARCA4|BAF 190|BAF190|BAF190A|Brahma protein like 1|BRG1|BRG1 associated factor 190A|BRG1 protein|BRG1-associated factor 190A|BRM/SWI2 related gene 1|Global transcription activator homologous sequence|global transcription activator snf2l4|Homeotic gene regulator|hSNF2b|Mitotic growth and transcription activator|MRD16|Nuclear protein GRB1|Protein brahma homolog 1|Protein BRG-1|Protein BRG1|RTPS2|SMARC A4|SMARCA4|SMCA4_HUMAN|SNF2|SNF2 beta|SNF2 like 4|SNF2-beta|SNF2B|SNF2L4|SNF2LB|Sucrose nonfermenting like 4|SWI/SNF related matrix associated actin dependent regulator of chromatin subfamily A member 4|SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily a, member 4|SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily A member 4|SWI2|Transcription activator BRG1
- ATP dependent helicase SMARCA4|ATP-dependent helicase SMARCA4|BAF 190|BAF190|BAF190A|Brahma protein like 1|BRG1|BRG1 associated factor 190A|BRG1 protein|BRG1-associated factor 190A|BRM/SWI2 related gene 1|Global transcription activator homologous sequence|global transcription activator snf2l4|Homeotic gene regulator|hSNF2b|Mitotic growth and transcription activator|MRD16|Nuclear protein GRB1|Protein brahma homolog 1|Protein BRG-1|Protein BRG1|RTPS2|SMARC A4|SMARCA4|SMCA4_HUMAN|SNF2|SNF2 beta|SNF2 like 4|SNF2-beta|SNF2B|SNF2L4|SNF2LB|Sucrose nonfermenting like 4|SWI/SNF related matrix associated actin dependent regulator of chromatin subfamily A member 4|SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily a, member 4|SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily A member 4|SWI2|Transcription activator BRG1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P51532
- P51532
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together