Anti-VIPR2 Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: VIPR2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77804-375
- 77804-374
- Antigen symbol:
- VIPR2
- VIPR2
- Antigen name:
- Vasoactive intestinal polypeptide receptor 2 protein (24-126AA)
- Vasoactive intestinal polypeptide receptor 2 protein (24-126AA)
- Antigen synonyms:
- Helodermin-preferring VIP receptor|PACAP type III receptor|PACAP-R-3|PACAP-R3|Pituitary adenylate cyclase-activating polypeptide type III receptor|Vasoactive intestinal polypeptide receptor 2|VIP Receptor 2|VIP-R-2|VIPR2_HUMAN|VPAC2 receptor
- Helodermin-preferring VIP receptor|PACAP type III receptor|PACAP-R-3|PACAP-R3|Pituitary adenylate cyclase-activating polypeptide type III receptor|Vasoactive intestinal polypeptide receptor 2|VIP Receptor 2|VIP-R-2|VIPR2_HUMAN|VPAC2 receptor
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P41587
- P41587
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together