Anti-TRIM23 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: WB: 1:1000 to 1:5000, IHC: 1:20 to 1:200
Type:
Antigen: TRIM23
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 77811-530
- 77803-530
- Antigen symbol:
- TRIM23
- TRIM23
- Antigen name:
- E3 ubiquitin-protein ligase TRIM23 protein (1-280AA)
- E3 ubiquitin-protein ligase TRIM23 protein (1-280AA)
- Antigen synonyms:
- ADP ribosylation factor domain protein 1|ADP-ribosylation factor domain-containing protein 1|ARF domain protein 1|ARFD1|E3 ubiquitin-protein ligase TRIM23|GTP binding protein ARD 1|GTP-binding protein ARD-1|N-acetyltransferase ARD1 human homolog of|N-acetyltransferase homolog of S. cerevisiae ARD1|RING finger protein 46|TRI23_HUMAN|TRIM23|Tripartite motif protein TRIM23|Tripartite motif-containing protein 23
- ADP ribosylation factor domain protein 1|ADP-ribosylation factor domain-containing protein 1|ARF domain protein 1|ARFD1|E3 ubiquitin-protein ligase TRIM23|GTP binding protein ARD 1|GTP-binding protein ARD-1|N-acetyltransferase ARD1 human homolog of|N-acetyltransferase homolog of S. cerevisiae ARD1|RING finger protein 46|TRI23_HUMAN|TRIM23|Tripartite motif protein TRIM23|Tripartite motif-containing protein 23
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P36406
- P36406
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together