Anti-ZWINT Rabbit Recombinant Polyclonal Antibody
New Product
Recommended dilution: WB: 1:1000 to 1:5000, IHC: 1:20 to 1:200
Type:
Antigen: ZWINT
Clonality: Recombinant Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
- Catalog No:
- 77819-426
- 77819-427
- Antigen symbol:
- ZWINT
- ZWINT
- Antigen name:
- ZW10 interactor protein (1-277AA)
- ZW10 interactor protein (1-277AA)
- Antigen synonyms:
- Human ZW10 interacting protein 1| HZwint 1| HZwint1| KNTC 2 AP| KNTC2AP| MGC 117174| MGC117174| ZW10 interacting kinetochore protein| ZW10 interacting protein 1| ZW10 interactor| kinetochore protein| ZW10-interacting protein 1| ZWINT 1| ZWINT| Zwint-1| ZWINT_HUMAN| ZWINT1
- Human ZW10 interacting protein 1| HZwint 1| HZwint1| KNTC 2 AP| KNTC2AP| MGC 117174| MGC117174| ZW10 interacting kinetochore protein| ZW10 interacting protein 1| ZW10 interactor| kinetochore protein| ZW10-interacting protein 1| ZWINT 1| ZWINT| Zwint-1| ZWINT_HUMAN| ZWINT1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O95229
- O95229
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together