Anti-IRF9 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:1000 to 1:2000, WB: 1:200 to 1:1000
Type:
Antigen: IRF9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Specifications
- Catalog No:
- 77818-365
- 77818-364
- Antigen symbol:
- IRF9
- IRF9
- Antigen name:
- Fusion protein of IRF9
- Fusion protein of IRF9
- Antigen synonyms:
- IFN alpha responsive transcription factor subunit| IFN-alpha-responsive transcription factor subunit| Interferon regulatory factor 9| interferon stimulated transcription factor 3| Interferon-stimulated gene factor 3 gamma| interferon-stimulated transcription factor 3, gamma 48kDa| IRF 9| IRF-9| Irf9| IRF9_HUMAN| ISGF 3 gamma| ISGF-3 gamma| ISGF3| ISGF3 p48 subunit| ISGF3G| OTTHUMP00000164692| OTTHUMP00000164693| p48| Transcriptional regulator ISGF3 subunit gamma
- IFN alpha responsive transcription factor subunit| IFN-alpha-responsive transcription factor subunit| Interferon regulatory factor 9| interferon stimulated transcription factor 3| Interferon-stimulated gene factor 3 gamma| interferon-stimulated transcription factor 3, gamma 48kDa| IRF 9| IRF-9| Irf9| IRF9_HUMAN| ISGF 3 gamma| ISGF-3 gamma| ISGF3| ISGF3 p48 subunit| ISGF3G| OTTHUMP00000164692| OTTHUMP00000164693| p48| Transcriptional regulator ISGF3 subunit gamma
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q00978
- Q00978
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together