Anti-ZFP36L2 Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: ZFP36L2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77804-937
- 77804-936
- Antigen symbol:
- ZFP36L2
- ZFP36L2
- Antigen name:
- mRNA decay activator protein ZFP36L2 protein (219-321AA)
- mRNA decay activator protein ZFP36L2 protein (219-321AA)
- Antigen synonyms:
- BRF2|Butyrate response factor 2|C3H1 type-like 2|EGF response factor 2|EGF-response factor 2|ERF 2|ERF-2|ERF2|Protein TIS11D|RNF162C|TIS11D|TISD_HUMAN|ZFP36 like 2|ZFP36 ring finger protein like 2|ZFP36-like 2|Zfp36l2|Zinc finger protein 36|Zinc finger protein 36, C3H type like 1|zinc finger protein 36, C3H type like 2|Zinc finger protein 36, C3H1 type like 2|Zinc finger protein, C3H type, 36 like 2
- BRF2|Butyrate response factor 2|C3H1 type-like 2|EGF response factor 2|EGF-response factor 2|ERF 2|ERF-2|ERF2|Protein TIS11D|RNF162C|TIS11D|TISD_HUMAN|ZFP36 like 2|ZFP36 ring finger protein like 2|ZFP36-like 2|Zfp36l2|Zinc finger protein 36|Zinc finger protein 36, C3H type like 1|zinc finger protein 36, C3H type like 2|Zinc finger protein 36, C3H1 type like 2|Zinc finger protein, C3H type, 36 like 2
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P47974
- P47974
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together