Anti-AIMP1 Rabbit Polyclonal Antibody
Catalog # 77798-890
Supplier: AFG Bioscience
New Product
Specifications
- Antigen name:SCYE1
- Antigen symbol:AIMP1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q12904
- Antigen synonyms:AIMP 1|Aimp1|AIMP1/p43|AIMP1_HUMAN|Aminoacyl tRNA synthetase complex-interacting multifunctional protein 1|ARS interacting multifunctional protein 1|EMAP 2|EMAP-2|EMAP-II|EMAP2|EMAPII|Endothelial monocyte activating polypeptide 2|Endothelial monocyte activating polypeptide|Endothelial monocyte-activating polypeptide 2|Endothelial monocyte-activating polypeptide II|HLD3|Multisynthase complex auxiliary component p43|Multisynthetase complex auxiliary component p43|OTTHUMP00000219673|p43|Scye 1|Scye1|Scye1, formerly|Small inducible cytokine subfamily E member 1|Small inducible cytokine subfamily E, member 1 (endothelial monocyte-activating)|Small inducible cytokine subfamily E, member 1, formerly
- Storage buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: AIMP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat