Anti-MRPS18A Rabbit Polyclonal Antibody
Catalog # 77806-308
Supplier: AFG Bioscience
New Product
Specifications
- Antigen name:Synthesized peptide derived from the Internal region of MRP-S18A.
- Antigen symbol:MRPS18A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Size:50 µg
- Form:Liquid
- Gene ID:Q9NVS2
- Antigen synonyms:MRPS18A|39S ribosomal protein S18a|mitochondrial|MRP-S18-a|Mrps18a|S18mt-a|39S ribosomal protein S18-3|MRP-S18-3|Mitochondrial large ribosomal subunit protein bS18a|Mitochondrial large ribosomal subunit protein mL66
- Storage buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on Blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: IHC: 1:100 to 1:300, ELISA: 1:40000
Type:
Antigen: MRPS18A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat