Anti-CDCA8 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: CDCA8
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77827-422
- 77827-423
- Antigen symbol:
- CDCA8
- CDCA8
- Antigen name:
- Borealin protein (1-280AA)
- Borealin protein (1-280AA)
- Antigen synonyms:
- BOR|BOREA_HUMAN|Borealin|CDCA8|Cell division cycle associated 8|cell division cycle associated protein 8|Cell division cycle-associated protein 8|Dasra B|Dasra-B|DasraB|FLJ10468|FLJ12042|hDasra B|hDasra-B|MESRGP|OTTHUMP00000004514|pluripotent embryonic stem cell related gene 3 protein|Pluripotent embryonic stem cell-related gene 3 protein
- BOR|BOREA_HUMAN|Borealin|CDCA8|Cell division cycle associated 8|cell division cycle associated protein 8|Cell division cycle-associated protein 8|Dasra B|Dasra-B|DasraB|FLJ10468|FLJ12042|hDasra B|hDasra-B|MESRGP|OTTHUMP00000004514|pluripotent embryonic stem cell related gene 3 protein|Pluripotent embryonic stem cell-related gene 3 protein
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q53HL2
- Q53HL2
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together